Yttrium aluminum ≤1% ≥99.9% (base métaux sauf Ca), en morceaux
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Purity: 99.9%
Molecular Formula: Y:Al; 87:12 wt%
MDL No.: MFCD01074900
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...