Aluminum phosphate ≥97%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
Aluminium phosphate
Purity: 97%
CAS No.: 7784-30-7
Molecular Formula: AlO4P
Formula Weight: 121.95
Physical Form: Powder
Density: 2.566
Appearance: White
Melting Point:
Applications: As a flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy
Formule:
AlPO₄ Poids moléculaire: 121.951 g/mol Densité: 2.566 Température de stockage: Température ambiante |
Numéro MDL:
MFCD00003430 Numéro CAS: 7784-30-7 EINECS: 232-056-9 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...