Imprimer
Hardness quality control standard
Marques : Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2833349 2833449
HACH2833349EA 110 EUR
HACH2833349 HACH2833449
Hardness quality control standard
Étalons Étalons pour l'analyse environnementale
These mixed standards are ready to use with no dilutions required. Includes standards for both total hardness and calcium hardness determination.