Imprimer
Kits de tests, cyanure, Spectroquant®, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.14561.0001 1.09701.0001
1.14561.0001EA 140 EUR
1.14561.0001 1.09701.0001
Kits de tests, cyanure, Spectroquant®, Supelco®
Kits de tests environnementaux
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Kits de test de réactifs Spectroquant®
-Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Remarque : la plage mentionnée dans le tableau ci-dessous se réfère aux instruments Prove et Nova. Pour certains kits de test, la plage de mesure disponible pour les instruments Spectroquant® Multy et Move 100 peut être réduite.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and waste water.