Imprimer
Quartz microfibre filters for air sampling, grade QM-B, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1852-042 1852-040
WHAT1852-042EA 206 EUR
WHAT1852-042 WHAT1852-040
Quartz microfibre filters for air sampling, grade QM-B, Whatman™
Filters Glass Fiber Filters
These high purity QM-B quartz fibre filters are designed for use in specific air monitoring applications.
QM-B filters are similar to QM-A quartz fibre filters, which are recommended more commonly for air testing applications. By comparison, QM-B is approximately twice as heavy, twice as thick and offers slower flow rates. Because of these characteristics, QM-B can be used in air monitoring applications requiring increased strength or durability.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Weight 170 g/m²
Thickness 950 µm
Airflow rate 12 s/100 ml/6,5 cm²
Max. temperature 800 °C