Imprimer
Hand cleansing paste, Solopol® natural
Fournisseur: Deb-STOKO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
33456 33282
EVON33456EA 142 EUR
EVON33456 EVON33282
Hand cleansing paste, Solopol® natural
Soin de la peau Savons et soins nettoyants
Solvent-free hand cleanser paste with scrubbing-agent for extra heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in general industrial environments.
  • Astopon® natural scrubbing agent gently removes contaminants without damaging the skin
  • Contains skin protectant Eucornol® and aloe vera
  • Formulation leaves hands feeling smooth and conditioned
  • Solvent-free, pH neutral to the skin
Certifications: EU Ecolabel
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904