Imprimer
Universal container with screw cap, PS, 30 ml, STERILIN®
Fournisseur: Sterilin
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
128SA 128SC 128SB 128C/FS 128A/FS 128B/FS
215-0325EA 387 EUR
215-0325 215-0327 215-0326 216-1043 216-1041 216-1042
Universal container with screw cap, PS, 30 ml, STERILIN®
Récipients de collecte d'échantillon Conteneurs à usage universel
Polystyrene, clear, with polypropylene screw cap.
  • Aseptically manufactured
  • 95 kPa-compliance ensures leak-free performance and suitability for transport (except spoon variants)
  • Available pre-filled with boric acid (0,4 g) for the preservation of urine samples
  • Suitable for centrifugation at 3800×g
  • In vitro use only
Ensure excellent sample containment using universal containers, featuring a traditional flow seal with 1-1/2 turn cap.
Certifications: Leak-tested in accordance with BS EN 14254 Annexe D. CE marked in accordance with the European Directive 98/79/EC.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904