Imprimer
Quickstart universal containers, PP, 30 ml, STERILIN®
Fournisseur: Sterilin
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
128A/P 128BBAC/P 128B/P 128C/P
216-1395EA 288 EUR
216-1395 216-1397 216-1396 216-1398
Quickstart universal containers, PP, 30 ml, STERILIN®
Récipients de collecte d'échantillon Conteneurs à usage universel
Polypropylene, ultra clear, with QickStart cap.
  • Aseptically manufactured
  • Suitable for centrifugation at 9500 ×g
  • Multi-seal cap 'QuickStart' for improved handling and unrivaled leak-free performance
  • Lot number printed on each container to aid full traceability
  • Labeled for patient details and available either undosed, dosed with boric acid preservative (0,4 g) or a loose spoon
Secure excellent temperature and chemical resistance with these 95kPa compliant and CE marked leak-resistant universal containers.
Certifications: Leak-tested in accordance with BS EN 14254 Annexe. CE marked in accordance with the European Directive 98/79/EC.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904