Imprimer
GC guard columns, HydroGOLD
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26H50-0232 26H50-0253 26H50-0525 26H50-0532 26H50-0553 26H50-1032 26H50-1053
555-9124EA 78.4 EUR
555-9124 555-9123 555-9121 555-9120 555-9119 555-9116 555-9115
GC guard columns, HydroGOLD
Colonnes pour chromatographie
These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.
  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • Protects against column contamination caused by non-volatile materials