Imprimer
Reagents for photometers
Marques : Palintest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AP011 AP155 AP031 AP041 AP152 AP156 AP177
PALIAP011EA 65.8 EUR
PALIAP011 PALIAP155 PALIAP031 PALIAP041 PALIAP152 PALIAP156 PALIAP177
Reagents for photometers
Kits de tests environnementaux
Photometer tablet reagents are ideal format for the testing of drinking water, natural water, wastewater and other process applications.
  • Portable, compact and stable
  • Simple test protocols - crush one or two tablets in the sample to carry out reliable water analysis
  • Wide range of parameters