Imprimer
Cover glasses for automated Shandon equipment, Menzel Gläser
Fournisseur: Epredia
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BB024050S113 BB024040SC13 BB024050SC13 BB024040S113 BB024060S113 BB024060SC13
631-0901EA 107 EUR
631-0901 630-2602 630-2603 631-0974 631-0973 630-2108
Cover glasses for automated Shandon equipment, Menzel Gläser
Lamelles couvre-objet
The cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness: Pure white glass of hydrolytic class 1
  • No bubbles or flaws
  • Pre-washed