Imprimer
Fittings, Uniflex™
Fournisseur: BROEN-LAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
25330591036-09 25330111036-09
BROE25330591036-09EA 1180 EUR
BROE25330591036-09 BROE25330111036-09
Fittings, Uniflex™
Faucets and Sinks Faucets
UniFlex™ BASIC fittings for special water are designed to secure maximum purity of the water in laboratory. The operation and configuration of the fittings are highly adapted to the requirements of a modern laboratory. These fittings are surface treated with polycoat powder coating, which is highly resistant to most chemicals, UV light and heat. Body made from LLDPE, POM, PP, PVDF / EPDM rubber.
  • Safe and simple installation
  • Ideal for corrosive media
  • Eliminate corrosion problems in connection with use of purified water
  • Spout with full swivel action
  • With removable plastic hose nozzle
  • Without push-in fitting
Max. working pressure: 10 bar at 20 °C; 6 bar at 65 °C.
Informations de livraison: Supplied with plastic handle and LLDPE (Linear LowDensity Polyethylene) inner pipe.