Imprimer
Leather gloves, classic driver, A270
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A270TARXL A270TARL
PRTWA270TARXLEA 116 EUR
PRTWA270TARXL PRTWA270TARL
Leather gloves, classic driver, A270
Gants
These tough, hard wearing gloves are made from premium full grain cow leather which provides a longer lasting glove.
  • Increased puncture resistance properties
  • Breathable leather for use in mild and hot climates
  • Superb abrasion and tear resistance
Certifications: EN 388:2016 (3142X).