Imprimer
Safety shoes, lace-up, The Roar, Jade
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SN20086/39 SN20086/40 SN20086/41 SN20086/42 SN20086/43 SN20086/44 SN20086/45 SN20086/46 SN20086/47 SN20086/48 SN20086/38
UPOWSN20086/39EA 99.2 EUR
UPOWSN20086/39 UPOWSN20086/40 UPOWSN20086/41 UPOWSN20086/42 UPOWSN20086/43 UPOWSN20086/44 UPOWSN20086/45 UPOWSN20086/46 UPOWSN20086/47 UPOWSN20086/48 UPOWSN20086/38
Safety shoes, lace-up, The Roar, Jade
Chaussures et bottes
Safety shoes made from velours leather and a slip-resistant dual-density PU sole.
  • Light non-slip Action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Beige
Certifications: EN 20345:2011; S1P, SRC