Imprimer
ESD safety shoes, Velcro® fastening, Rock & Roll, Oak
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RR30306/35 RR30306/36 RR30306/37 RR30306/38 RR30306/39 RR30306/40 RR30306/41 RR30306/42 RR30306/43 RR30306/44 RR30306/45 RR30306/46 RR30306/47 RR30306/48
UPOWRR30306/35EA 93.9 EUR
UPOWRR30306/35 UPOWRR30306/36 UPOWRR30306/37 UPOWRR30306/38 UPOWRR30306/39 UPOWRR30306/40 UPOWRR30306/41 UPOWRR30306/42 UPOWRR30306/43 UPOWRR30306/44 UPOWRR30306/45 UPOWRR30306/46 UPOWRR30306/47 UPOWRR30306/48
ESD safety shoes, Velcro® fastening, Rock & Roll, Oak
Chaussures et bottes
100% metal-free ESD safety shoes made from smooth leather with abresion resistant toe protection and a slip-resistant dual-density PU sole.
  • Shock absorbing, anti fungal PE insole
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non metallic, puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certifications: EN 61340 (ESD); EN 20345:2011; S1P, SRC