Imprimer
Two-dimensional BioProcess Container™ system, Productainer™ BPC bags
Fournisseur: THERMO BIOPROCESS DIVISION
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30651.06 SH30651.07
HYCPSH30651.06EA 2390 EUR
HYCPSH30651.06 HYCPSH30651.07
Two-dimensional BioProcess Container™ system, Productainer™ BPC bags
Bags Media Bags
BioProcess Container™ (BPC™) systems include two- and three-dimensional single-use bags for packaging, storage and transport of 50 to 200 litre volumes of media. Manufactured from medical-grade plastics (cGMP compliant) and configured with ports, tubing and fittings, BPC™ systems provide an efficient means for sterile liquid handling in research and production settings.
  • Film type: CX3-9 (except Productainer™ BPC bag with 2 top and 1 bottom ports, CX5-14)
  • Sterilisation method: Gamma irradiation