Imprimer
Safety shoes, lace-up, Concept plus, Eruption
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UA20394/38 UA20394/39 UA20394/40 UA20394/41 UA20394/42 UA20394/43 UA20394/44 UA20394/45 UA20394/46 UA20394/47
UPOWUA20394/38EA 107 EUR
UPOWUA20394/38 UPOWUA20394/39 UPOWUA20394/40 UPOWUA20394/41 UPOWUA20394/42 UPOWUA20394/43 UPOWUA20394/44 UPOWUA20394/45 UPOWUA20394/46 UPOWUA20394/47
Safety shoes, lace-up, Concept plus, Eruption
Chaussures et bottes
Safety shoes made from water repellent full grain leather and a heat resistant to 300 °C, slip resistant nitrile rubber sole.
  • Light non-slip action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex'covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certifications: EN 20345:2011; S3, HI, CI, HRO, RS