Imprimer
Safety shoes, lace-up, Concept Plus, Elite
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UA20564/35 UA20564/36 UA20564/37 UA20564/38 UA20564/39 UA20564/40 UA20564/41 UA20564/42 UA20564/43 UA20564/44 UA20564/45 UA20564/46 UA20564/47 UA20564/48
UPOWUA20564/35EA 75.8 EUR
UPOWUA20564/35 UPOWUA20564/36 UPOWUA20564/37 UPOWUA20564/38 UPOWUA20564/39 UPOWUA20564/40 UPOWUA20564/41 UPOWUA20564/42 UPOWUA20564/43 UPOWUA20564/44 UPOWUA20564/45 UPOWUA20564/46 UPOWUA20564/47 UPOWUA20564/48
Safety shoes, lace-up, Concept Plus, Elite
Chaussures et bottes
100% metal-free safety shoes made from water repellent full grain leather and a hard wearing, slip resistant PU sole.
  • Light non-slip action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex'covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certifications: EN 20345:2011; S3, SRC