Imprimer
Safety shoes, lace-up, Concept Plus, Twister
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UC20036/38 UC20036/39 UC20036/40 UC20036/41 UC20036/42 UC20036/43 UC20036/44 UC20036/45 UC20036/46 UC20036/47 UC20036/48 UC20036/36 UC20036/37
UPOWUC20036/38EA 83.6 EUR
UPOWUC20036/38 UPOWUC20036/39 UPOWUC20036/40 UPOWUC20036/41 UPOWUC20036/42 UPOWUC20036/43 UPOWUC20036/44 UPOWUC20036/45 UPOWUC20036/46 UPOWUC20036/47 UPOWUC20036/48 UPOWUC20036/36 UPOWUC20036/37
Safety shoes, lace-up, Concept Plus, Twister
Chaussures et bottes
Metal-free safety shoes made from soft suede leather with highly breathable textile mesh inserts and a hard wearing, anti-static, antislip PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective composite toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Grey
Certifications: EN 20345:2011; S1P, SRC