Imprimer
Safety ankle boots, lace-up, SK Grip, Tweed Grip
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SK Grip Tweed Grip
UPOWUK10074/46EA 90.3 EUR
UPOWUK10074/46 UPOWUK10074/47 UPOWUK10074/38 UPOWUK10074/42 UPOWUK10074/43 UPOWUK10074/44 UPOWUK10074/45 UPOWUK10074/40 UPOWUK10074/41 UPOWUK10074/39
Safety ankle boots, lace-up, SK Grip, Tweed Grip
Chaussures et bottes
Safety ankle boots made from water repellent full grained leather and a lightweight, extremely flexible, slip resistant PU/PU U-Grip sole.
  • Anatomically shaped Polysoft insole made from soft PU, with pockets in the foot support area
  • PU/PU walking sole with 90° angle blocks and mulitlevel stabalising channels to for extremely high anti-slip features; TPU anti twist insert for additional support
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe aluminium protective toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certifications: EN 20345:2011; S3, SRC