Imprimer
Safety ankle boots, lace-up, Concept M, Track
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UM10013/36 UM10013/37 UM10013/38 UM10013/39 UM10013/40 UM10013/41 UM10013/42 UM10013/43 UM10013/44 UM10013/45 UM10013/46 UM10013/47 UM10013/48
UPOWUM10013/36EA 98 EUR
UPOWUM10013/36 UPOWUM10013/37 UPOWUM10013/38 UPOWUM10013/39 UPOWUM10013/40 UPOWUM10013/41 UPOWUM10013/42 UPOWUM10013/43 UPOWUM10013/44 UPOWUM10013/45 UPOWUM10013/46 UPOWUM10013/47 UPOWUM10013/48
Safety ankle boots, lace-up, Concept M, Track
Chaussures et bottes
Anti-static safety ankle boots made from water repellent full grain leather and a hard wearing, slip resistant PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Puncture resistant midsole made from steel
Colour: Black
Colour: Black
Certifications: EN 20345:2011; S3, SRC