Imprimer
Safety shoes, lace-up, Concept M, Mustang
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Concept M Mustang
UPOWUM20013/41EA 90.2 EUR
UPOWUM20013/41 UPOWUM20013/40 UPOWUM20013/43 UPOWUM20013/42 UPOWUM20013/45 UPOWUM20013/44 UPOWUM20013/47 UPOWUM20013/36 UPOWUM20013/46 UPOWUM20013/38 UPOWUM20013/37 UPOWUM20013/48 UPOWUM20013/39
Safety shoes, lace-up, Concept M, Mustang
Chaussures et bottes
Anti-static safety shoes made from water repellent full grain leather and a hardwearing, anti-slip PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective composite toecap with breathable membrane (200 J)
  • Classic steel midsole
Colour: Black
Certifications: EN 20345:2011; S3, SRC, HI, HRO