Imprimer
Safety shoes, lace-up, U-Gel, Caption
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UR20114/39 UR20114/38 UR20114/40 UR20114/41 UR20114/42 UR20114/43 UR20114/44 UR20114/45 UR20114/46 UR20114/47
UPOWUR20114/39EA 98.3 EUR
UPOWUR20114/39 UPOWUR20114/38 UPOWUR20114/40 UPOWUR20114/41 UPOWUR20114/42 UPOWUR20114/43 UPOWUR20114/44 UPOWUR20114/45 UPOWUR20114/46 UPOWUR20114/47
Safety shoes, lace-up, U-Gel, Caption
Chaussures et bottes
Safety shoes made from full grain leather with Hi Viz inserts water resistant and a shock absorbing TPU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certifications: EN 20345:2011; S3, SRC