Imprimer
Measuring cylinders , SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F286920000 F286910000 F286930000 F286950000 F286970000
BELAF286920000EA 32.7 EUR
BELAF286920000 BELAF286910000 BELAF286930000 BELAF286950000 BELAF286970000
Measuring cylinders , SP Bel-Art
Éprouvettes Éprouvettes graduées
PMP (TPX), glass-clear, tall form.
  • Shatter-proof
  • Wide octagonal base prevents tipping and rolling
  • Very good chemical resistance, temperature resistance to 175 °C
Tall form cylinders are non-wetting to prevent a concave meniscus, making the raised numeral, single scale graduations easy to read.