Imprimer
Safety shoes, lace-up, White 68 & Black, Rebound Grip
Fournisseur: U-Power
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
UW20022/35 UW20022/36 UW20022/37 UW20022/38 UW20022/39 UW20022/40 UW20022/41 UW20022/42 UW20022/43 UW20022/44 UW20022/45 UW20022/46 UW20022/47 UW20022/48
UPOWUW20022/35EA 69.3 EUR
UPOWUW20022/35 UPOWUW20022/36 UPOWUW20022/37 UPOWUW20022/38 UPOWUW20022/39 UPOWUW20022/40 UPOWUW20022/41 UPOWUW20022/42 UPOWUW20022/43 UPOWUW20022/44 UPOWUW20022/45 UPOWUW20022/46 UPOWUW20022/47 UPOWUW20022/48
Safety shoes, lace-up, White 68 & Black, Rebound Grip
Chaussures et bottes
100% metal-free safety shoes made from breathable and water repellent New Safety Dry material and a slip and abrasion resistant U.Grip68 PUR sole.
  • White anatomically shaped Ergodry insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • AirToe Composite toecap with breathable membrane (200 J)
Colour: White
Certifications: EN 20345:2011; S2, SRC