Imprimer
Magnetic stirring bars, polygon, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F371209020 F371209030 F371209040 F371209070 F371209080 F371220035
BELAF371209020EA 7.8 EUR
BELAF371209020 BELAF371209030 BELAF371209040 BELAF371209070 BELAF371209080 BELAF371220035
Magnetic stirring bars, polygon, Spinbar®, SP Bel-Art
Agitateurs Barreaux magnétiques
PTFE covered with strong Alnico V magnetic core. The multifaceted surfaces add turbulence relative to smooth cylindrical bars of a similar size.
  • Autoclave safe
  • FDA grade PTFE is durable and inert
  • Pivot ring minimises the contact area between the bar and the vessel