Imprimer
Pointes de pipettes, LTS®, pour pipettes Rainin
Fournisseur: RAININ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17001131 17001129 30389271 30389277 30389270 30389200 17001133 17001119 30389215 30389273 30389275 30389278 30389294 30389297 30389299 30389301 30389304 17000504 30389211 30389228 30389230 30389234 30389238 30389245 30389255 30389256 30389257 17007082 30389243
RAIN17001131EA 147 EUR
RAIN17001131 RAIN17001129 RAIN30389271 RAIN30389277 RAIN30389270 RAIN30389200 RAIN17001133 RAIN17001119 RAIN30389215 RAIN30389273 RAIN30389275 RAIN30389278 RAIN30389294 RAIN30389297 RAIN30389299 RAIN30389301 RAIN30389304 RAIN17000504 RAIN30389211 RAIN30389228 RAIN30389230 RAIN30389234 RAIN30389238 RAIN30389245 RAIN30389255 RAIN30389256 RAIN30389257 RAIN17007082 RAIN30389243
Pointes de pipettes, LTS®, pour pipettes Rainin
Pointes de pipettes
These thin-wall tips are designed for use with RAININ Pipet-Lite, Pipet-Plus and EDP3-Plus pipettes with LTS LiteTouch Tip Ejection System. 1200 µl tips are specifically designed for use with RAININ E12-1200 and E8-1200 multi-channel pipettes. The unique design prevents drop formation and eliminates the need for touch-off between dispenses. The tips are available in different packaging options.
  • Virgin PP, sterile or non sterile
  • Wide orifice tips (1,5 mm) minimise shear forces on delicate samples and flow resistance for viscous samples
  • Extended length tips (102 mm long) will reach into the bottom of blocks, tall vials, flasks, 100 mm tubes etc.
  • Low retention tips feature an ultra-hydrophobic tip surface that minimises liquid retention; ideal for pipetting proteins, enzymes and DNA
  • 20 ml tips have graduation marks
Bulk
- Packaged in zip-lock bags
- Very economical
Racks with removable cover
- Sealed bottoms and overlapping lids protect tips from dust, dirt and micro-organisms
- Two-position lid allows autoclaving with the lid in vent position
- Sturdy shelf construction allows easy tip loading with single and multi-channel pipettes
Green-Pak® Individual Refills and Green-Pak® SpaceSaver® Refills
- Environmentally friendly refills for racks; less plastic waste
- Individual refills contain 96 tips in 8×12 format, sealed in a shell of Code 1 PET recyclable plastic
- Each refill is completely sealed to protect the tips; hands and fingers never contact the tips during loading
- SpaceSaver® systems are provided with either eight or ten stacked refills
- SpaceSaver® refills are packaged in a code 1 PET sleeve which functions as an automatic tip dispenser that aligns and refills an empty rack with 96 tips at a time
StableStak™ racks
- 960 tips in five stacked double-96 racks
- Strong, flat trays for even tip loading
- Ideal for multi-channel pipetting in high-production environments
- Saves time, space and plastic waste
StableRak™
- 192 tips in one double-96 rack
- Each StableRak™ contains 192 tips in one rack instead of two