Imprimer
Immersion thermostats, IC series
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3861000 3863000
531-1061EA 4070 EUR
531-1061 531-1062
Immersion thermostats, IC series
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The IC immersion circulators have an extended temperature range for tempering fluids in open and covered water baths up to 200/250 °C. The LED/ graphic TFT Display illustrates all relevant process parameters. Units feature push and turn buttons for intuitive menu navigation. IC immersion circulators ensure short heating up times and have a temperature stability of ±0,01/0,02 K (basic/control model). Both models feature optical and acoustic safety warnings, an adjustable safety circuit, a powerful pressure/suction pump and can be used for internal and external applications.
  • A Wireless Controller (WiCo) enables safe operation of control models from a distance of up to 10 meters, e.g. in front of a closed fume hood (control device only)
  • Large easy-to-read LED display for pump speed and temperature on basic units, control units have a TFT display for temperature, pump speed as well as liquid level and safety temperature
  • The control models have timer function: 0 to 99 h 59 min, and a Mulit IO (Input/Output) interface to connect and control for e.g. electronic magnetic valves. Cooling coil is already included
  • USB/RS232 interface for PC control, documentation using labworldsoft® and for online firmware updates, as well as PT100 interface for external temperature control
  • The user can switch from internal to external temperature regulation with only one key using the control model
  • Integrated connections M16×1 for external applications
Max. flow rate/max pressure:
31 L/min, 610 mbar, (pressure), 450 mbar (suction)
* Lower working temperatures possible with connection to a chiller.
Certifications: Comply with DIN 12876, Safety class III, (FL) for use with flammable liquids. Units are designed in accordance to UL 61010-1.
Informations de livraison: Supplied with telescopic stainless steel bath-bridge for connection to water baths with different volumes. Both devices have a PT100 interface for external temperature control.