Imprimer
Casamino Acids, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0210477801 113060012 0296013801 0296013805 0210129101 0210129105 0210477805 0210477825 091010817 113060022
ICNA0210477801EA 216 EUR
ICNA0210477801 ICNA113060012 ICNA0296013801 ICNA0296013805 ICNA0210129101 ICNA0210129105 ICNA0210477805 ICNA0210477825 ICNA091010817 ICNA113060022
Casamino Acids, MP Biomedicals
Milieux pour microbiologie Matières premières de microbiologie
Casamino acids is supplemented to microbiological growth media.
  • Media Supplement
  • Bacteria Cell Culture
  • Base ingredient for media
Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.