Imprimer
VWR®, Ergonomic Laboratory Chairs and Stools
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
132-1099EA 364 EUR
132-1099 132-1100 132-1104 132-1105 132-1301 132-1300
VWR®, Ergonomic Laboratory Chairs and Stools
Chairs
Nobody has a perfectly symmetric morphology. This is why majority of people have back problems, sometimes with a feeling of heavy legs. The answer to this problem lies in the main features of this seat. Above all, a good working position is a dynamic posture that facilitates the bloodstream. The features of the seat help to re-balance the body naturally and improve blood flow in the legs. VWR® Ergonomic lab chairs and stools adapt automatically to the body and the movements of the user.
  • Automatically adapts to the user
  • Facilitates an upright posture
  • Minimises spinal stress
  • Seat height adjustment by gas lift
  • Black steel, 5-branch, scuff resistant reinforced base
  • Resistant against sparks, chemicals and weak acids
The innovative seat design consist of a short seat tilted forward, split into 2 parts, based on blue silent blocks.
The PU foam seat has a thickness of 40 mm and great deformation capacity which provides better distribution of pressure when moving.