Imprimer
Magnetic beads, Sera-Mag SpeedBeads Amine-blocked particles
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
19152104010150 19152104010350 19152104011150
19152104010150.EA 598 EUR
19152104010150. 19152104010350. 19152104011150.
Magnetic beads, Sera-Mag SpeedBeads Amine-blocked particles
Perles Billes magnétiques
Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.
  • Non-surfactant, non-protein blocking reagent provides very low non-specific binding of proteins from the sample matrix
  • Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
  • Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
  • Uniform, nominal 1 μm diameter provides excellent lot-to-lot reproducibility
Sera-Mag SpeedBeads Amine-Blocked Particles combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics. Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays.