Imprimer
Latex gloves, Rotiprotect®
Fournisseur: Roth Carl
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5884.1 5885.1 5886.1
112-0043EA 0 EUR
112-0043 112-0044 112-0045
Latex gloves, Rotiprotect®
Gants
These disposable gloves are suitable for protection against complex risks and for food processing.
  • Micro coarse outer surface
  • Smooth inner surface, lightly powdered
  • Very smooth and pleasantly soft
  • Thin rolled edge
  • Thiuram-free
Certifications: EN 374; Cat. III; AQL 1,5