Imprimer
Anemometers, smart and wireless probes, testo 405i/410iand VAC kit operated with smartphone
Fournisseur: TESTO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0560.1405 0560.1410
620-2395EA 153 EUR
620-2395 620-2396
Anemometers, smart and wireless probes, testo 405i/410iand VAC kit operated with smartphone
Anénomètres
These Smart Probes are designed to be used with smartphones and tablets. Users can read the measurements using the testo Smart Probes App installed on their smart device. The App allows you to remotely read measurements from up to six Smart Probes and easily document/report the results. All measurement data is displayed as instrument readings, tables, or graphs. The measurements can be quickly saved as PDF or Excel files. The App creates custom reports which can be saved and/or shared by email. The app works on smart devices with either Android or Apple operating systems (requires iOS 8.3 or newer; requires Android 4.3 or newer; requires mobile end device with Bluetooth 4.0.).
testo 405i - Hot-wire anemometer:
Wireless instrument for measuring air flow velocity, temperature, and volume flow
Measures in-duct air flow and automatically calculates volume flow (enter duct dimensions simply)
Small diameter (<0,5") and 15" telescopic shaft length is ideal for in-duct flow averaging and timed traverse
Perform air duct leakage tests
testo 410i - Vane anemometer:
Wireless instrument for measuring air velocity, volume flow, and temperature as well as air flow at registers
Automatically calculates volume flow (enter duct dimensions simply)
No air density correction required due to design
Measure volume flow at several outlets simultaneously (using mutiple Smart Probes) to adjust system balance
Timed/multipoint averaging to get accurate air flow profiles
testo VAC kit:
testo Smart Probes VAC kit for servicing on ventilation and air conditioning systems. For the measurement of air and surface temperature, humidity, air velocity and volume flow.
Informations de livraison: testo 405i includes Bluetooth® hot-wire anemometer with App, 3 AAA batteries and certificate of calibration.
testo 410i includes Bluetooth® vane anemometer with App, 3 AAA batteries and certificate of calibration.
testo VAC kit includes testo 405i, testo 410i, testo 605i (thermo-hygrometer), testo 805i (infrared thermometer), Smart Case, batteries, and certificate of calibration.