Imprimer
Safety dispensing cans, HDPE
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
14002 14004 14010
JUSU14002EA 58 EUR
JUSU14002 JUSU14004 119-0275
Safety dispensing cans, HDPE
Bidons de sécurité
These FM approved brass valves are ideal for use with flammables.
HDPE, textured.
  • Permit easy, one-hand dispensing of liquids directly onto work surface
  • Leakproof, self-closing, brass dispenser valve minimises spills and controls excess