Imprimer
Safety cans, type I, galvanised steel
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7150100Z 7125100Z 7120100Z
119-0413EA 65 EUR
119-0413 119-0411 119-0409
Safety cans, type I, galvanised steel
Bidons de sécurité
Safety cans, designed with a single spout for filling and pouring, the sealed lid features automatic positive-pressure relief that vents between 3 and 5 psig (0,2 and 0,35 bar) to guard against rupture or explosion.
  • High-gloss powder paint finish resists solvents, corrosion and humidity
  • Rounded steel comfort handle protects hand injury
  • Trilingual warning label in English/Spanish/French
  • Large ID zone identifies contents to reduce misuse
Made from high-grade coated steel, each can is tested to guarantee 100% leakproof construction. The tough powder-coat finish resists chemicals and stands up to demands of rigorous use. Each Justrite safety can is equipped with a stainless steel flame arrester which dissipates heat to stop flashback ignition. Cans up to 1 quart (1 L) feature a trigger-release handle, while larger capacities have a swinging handle to greatly ease carrying of heavier loads. Large label ID zone on swing-handle cans provides writing space to identity contents, department location, or individual user name.
Certified by FM, UL/ULC, and TÜV, Justrite safety cans also meet OSHA and NFPA requirements, and come with a ten-year warranty. - See more at: http://www.justritemfg.com/products/safety-cans-and-containers/type-i-steel-safety-cans/type-i-safety-cans-for-flammables/type-i-steel-safety-can-for-flammables-with-trigger-handle-1-gallon-ss-flame-arrester-self-close-lid11/#sthash.CZgZU7gh.dpuf