Imprimer
Spill containment caddy, EcoPolyBlend™
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28665 28664
119-0386EA 1560 EUR
119-0386 119-0385
Spill containment caddy, EcoPolyBlend™
Stockage des fûts
Made up of 90% recycled polyethylene.
  • Single-piece, double-walled seamless design eliminates leaks
  • Ergonomic design with tilt/stop feature minimises back strain
  • Rubber, 254 mm wheels roll over bumpy terrain use indoors or out
They offer convenient handling and dispensing of hazardous chemicals with complete assurance of compliance with EPA and SPCC regulations. It is the ideal choice for managing either 110 or 200 litre drums and the nylon strap holds drum securely in place.