Imprimer
Scrapers, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F368310000 F368300000 F368320001 F368310001
BELAF368310000EA 385 EUR
BELAF368310000 BELAF368300000 BELAF368320001 BELAF368310001
Scrapers, SP Bel-Art
Raclettes
HDPE or LDPE. These smooth plastic scrapers are ideal for removing residues from filters, cleaning containers, mixing and distributing solids in fluids, spreading pastes over large areas, and many other scraping, spreading, stirring or residue removing tasks.
  • Available in soft, FDA grade LDPE or in hard HDPE
  • Chemical resistant
  • Blade dimensions: 127×152 mm
  • Tapers from 9,5 mm down to a chisel edge
  • Upper end extends into a hexagonal shank 35 mm wide
Length of the handle: 250, 914 or 1830 mm