Imprimer
Safety Funnel, for drums
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
08207 08205 08227
119-0232EA 489 EUR
119-0232 119-0231 119-0233
Safety Funnel, for drums
Funnels Safety Funnels
Galvanised steel, 18-gauge, with electrostatically applied epoxy powder coat paint.
  • Wide mouth opening, stay-opened lid
  • Self-closing lin incase of fire
  • Built-in flame arrester on most models to prevent fire intrusion
  • Funnels for use with viscous liquids are fitted with brass tube
These safety drum funnels are mainly used to prevent spills while transferring flammable solvents, thinners, oils or paint wastes.
Approved by FM and compliance with EPA and OSHA regulations.