Imprimer
Culture tubes
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SCT-18100/TOC 3115-OTWS-2 CT18-TOC CT18-TOC/LL
EPSCSCT-18100/TOCEA 1460 EUR
EPSCSCT-18100/TOC EPSC3115-OTWS-2 EPSCCT18-TOC EPSCCT18-TOC/LL
Culture tubes
Tubes Tubes de culture
Culture tubes are designed for the Total Organic Carbon (TOC) testing and sampling.
  • USP method 643 testing
  • Off-line and grab sampling of high purity water
  • Low background is perfect for preparation and storage of standards
  • Cleaning validation is simplified and costs are reduced by using the TOC method of testing
All containers have open-top caps, unless otherwise specified, and fluoropolymer resin-lined septa.
Manufactured, tested and packaged by exacting standards, Thermo Scientific™ TOC Certified Containers are certified to contribute <10 ppb or <20 ppb TOC as background.
Vials feature a septum designed to stay inside the cap while sampling and are appropriate for use as autosampler vials or for sampling.
Informations de commande: Supplied with Certificate of Analysis.