Imprimer
Bottles, total organic carbon (TOC)
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S114-125A/TOC 1000A/TOC S114-250A/TOC S114-250C/TOC
EPSCS114-125A/TOCEA 303 EUR
EPSCS114-125A/TOC EPSC1000A/TOC EPSCS114-250A/TOC EPSCS114-250C/TOC
Bottles, total organic carbon (TOC)
Flacons Flacons en verre
Bouston bottles are designed for the Total Organic Carbon (TOC) testing and sampling.
  • USP method 643 testing
  • Off-line and grab sampling of high purity water
  • Low background is perfect for preparation and storage of standards
  • Cleaning validation is simplified and costs are reduced by using the TOC method of testing