Imprimer
Trimming blades and handles, FEATHER®
Fournisseur: PFM Medical GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
205500000 205570000 205540001 205530001 205550001 205500001 205500000_U 205560000
FEAT205500000EA 170 EUR
FEAT205500000 FEAT205570000 FEAT205540001 FEAT205530001 FEAT205550001 FEAT205500001 FEAT205500000_U FEAT205560000
Trimming blades and handles, FEATHER®
Microtomes and Microtome Assemblies Microtome Blades and Handles
The carbon steel trimming blades are special blades for cutting tissue samples without causing deformations. The trimming blade handles are made from heat-resistant plastic and due to their ergonomic design, they are particularly easy to grip.
  • Super sharp blades
  • Securely-fitted blade can be easily attached and removed