Imprimer
Avantor® ACE®, UHPLC Column Connectors
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
EXL-TWEA 924 EUR
EXL-TW EXL-CCSK EXL-CC10 EXL-CC
Avantor® ACE®, UHPLC Column Connectors
Raccords et connecteurs
Stainless steel/PEEK UHPLC column inlet / outlet connectors. These column connectors are designed to connect UHPLC columns to UHPLC systems and make other similar connections. They are suitable for use with ¹/₁₆" ext.Ø tubing and virtually all manufacturers' UHPLC systems and all brands of UHPLC columns.
  • Pressure rated to 1700 bar (>25000 psi)
  • Usable at elevated temperatures (up to 100 °C)
  • Reusable, non-permanent swaging design
  • 10 make and break cycles
For optimum connector lifetime and to avoid accidental permanent swaging of the connector to the tubing by over tightening, the use of an ACE® torque wrench is recommended.