Imprimer
Conductivity sensors
Fournisseur: Mettler - Toledo
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
51340335 30266974
663-0130EA 241 EUR
663-0130 661-0248
Conductivity sensors
Électrodes Électrodes de conductivité
This conductivity measuring cell utilises the four- pole potentiometric method for measuring conductivity, which incorporates a series of four graphite poles embedded in the probe shaft. This design completely eliminates the polarisation, which frequently occurs with the two-plate amperometric method during measuring high conductivity solution.
  • Four cell graphite sensor
  • Ideal sensor for general conductivity measurements
  • NTC 30 kΩ combined temperature sensor: automatic temperature compensation
Please note: Not recommended for samples below 10 μS/cm.
Informations de livraison: Supplied with quality certificate.