Imprimer
1000 and 1700 series Gastight® syringes for PSD/4 syringe pumps
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5495-50 5495-10 5495-15 5495-20 5495-25 5495-30 5495-35 5495-40 5495-45 5495-55 5495-22 5495-38
HAMI5495-50EA 245 EUR
HAMI5495-50 HAMI5495-10 HAMI5495-15 HAMI5495-20 HAMI5495-25 HAMI5495-30 HAMI5495-35 HAMI5495-40 HAMI5495-45 HAMI5495-55 HAMI5495-22 HAMI5495-38
1000 and 1700 series Gastight® syringes for PSD/4 syringe pumps
Seringues Seringues de chromatographie
Syringes for the PSD/4 syringe pump are available with a standard PTFE tipped plunger and with a long life UHMWPE (Ultra High Molecular Weight Poly Ethylene) plunger tip. The stroke length of a PSD/4 syringe is 30 mm.
Syringe terminations:
TLL, PTFE® Luer Lock
This termination has a PTFE, male Luer taper with nickel plated brass locking hub for use with Kel-F needles, metal hub needles and universal connectors. Also, the TLL is used with Hamilton Diluters/Dispensers, OEM applications and manual operations. Autoclavable when disassembled, except on 25 ml and greater syringes. Repeated autoclaving will shorten syringe life.
Plunger types:
X-style
This plunger is used for syringes 500 µl and smaller intended for use in an automated syringe pump. A special plunger button protects the delicate plunger wire by stopping the pump at the zero line before damaging the tip. The stop also provides a 6-32 threaded hole that is used to mount the plunger into the syringe drive.