Imprimer
Baths and circulators, stainless steel, CORIO™ C-B and CD-B series
Fournisseur: JULABO GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9012419 9011405 9011413 9011417 9011419 9011427 9012405 9012413 9012417 9012427 9012433 9012439 9012417.02
461-0912EA 2090 EUR
461-0912 461-0904 461-0905 461-0906 461-0907 461-0908 461-0909 461-0910 461-0911 461-0913 461-0914 461-0915 JULA9012417.02
Baths and circulators, stainless steel, CORIO™ C-B and CD-B series
Thermostats à circulation Systèmes à circulation ouverte Bains avec circulateurs détachables
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORIO™ C immersion circulator is the basic model, it is ideal for standard internal applications, the CD model is more advanced and can used for temperature control of external applications if required. The jet nozzle allows continuous adjustment of the pump stream in the bath. The stainless steel baths are durable and have an integrated drain tap (except B5). These open heating bath circulators are ideal for temperature control of samples, preparation of samples for serology and clinical chemistry, analysis, and material testing. The CD-B units are ideal for external temperature control applications in combination with measuring instruments, measuring cells, photometers, refractometers and polarimeters.
  • Precise temperature control
  • Bright, white, easy to read display
  • Very quiet and easy operation
  • CD version has a USB interface and pump can change between internal and external circulation via a lever located directly below the display
  • Ergonomic baths have a cleverly integrated drip-off design
Max. flow rate / max. pressure:
C: 6 L/min, 0,1 bar
CD: 15 L/min, 0,35 bar.
Certifications: C models: Class I (NFL), CD models: Class III (FL) according to DIN 12876-1.
Informations de livraison: Supplied with bath. CD models include 2 each of 8/12 mm Ø barbed fittings. A wide range of optional accessories, such racks, bath lids, tubing and adapters are available.