Imprimer
Pipette tips, FlexiBulk, Optifit
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LH-B790204 LH-B790354 LH-B791004 LH-B791024 LH-B791204
613-3887EA 44.2 EUR
613-3887 613-3888 613-3889 613-3890 613-3891
Pipette tips, FlexiBulk, Optifit
Pointes de pipettes
The FlexiBulk concept combines the low cost of bulk tips with saving space, improving usability and securing tip purity.
  • Reduces up to 40% on pack volume; cuts packaging waste by nearly 50%
  • Saves time and effort: Well-organized FlexiBulk pack makes tip picking quicker and more convenient
  • Tips are certified as free of RNase, DNase and endotoxins
  • Tips are manufactured under ISO Class 8 cleanroom conditions
  • Lid provides an airtight seal during transport and storage for secured purity
  • Lid is easy to close between uses to protect the tips from dust
FlexiBulk is small-sized and made of durable PET (polyethylene terephthalate) material that can be used and stacked as is – without any outer cardboard box needed. The orderly arrangement of tips allows the FlexiBulk tip packs to be approximately 40% smaller than conventional bulk tip cartons, where tips are packed in random order.
Certifications: Compliance with ISO 9001, ISO 14001 and ISO 13485 quality standards.