Imprimer
pH combination electrodes, IDS®
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285100080 285100120 285101080
SCOI285100080EA 780 EUR
SCOI285100080 661-0244 SCOI285101080
pH combination electrodes, IDS®
Électrodes Électrodes pH
When digital IDS sensors are connected to the measuring device, they automatically identify themselves with their serial numbers and type designation and transmit their calibration data to the device.
  • Calibration data is stored in the sensor head
  • Measuring signals are transmitted without interference
  • Perfect galvanized separation
  • Resistant against environmental influences
  • Allows prognostic maintenance of the sensors by the intelligent sensor evaluation QSC
  • Effortless allocation and documentation of the sensor to electronically captured and saved calibration results
  • Highest possible operator comfort and measuring accuracy