Imprimer
Syringes for thin layer chromatography
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
80050 80250 80950 81050 CAL81050 CAL80050
HAMI80050EA 115 EUR
HAMI80050 HAMI80250 HAMI80950 HAMI81050 HAMICAL81050 HAMICAL80050
Syringes for thin layer chromatography
Seringues Seringues de chromatographie
The needle points of these thin layer chromatography syringes are covered with a PTFE coating which reduces the surface tension between the needle and the liquid making it ideal for reproducible sample spotting.