Imprimer
Salt bridges
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B40A520 B40A610 B40A550
HACHB40A520EA 73.6 EUR
HACHB40A520 HACHB40A610 HACHB40A550
Salt bridges
Tubes Tubes à essais
During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.