Imprimer
Kjeldahl distillation systems, VAPODEST® 400 and VAPODEST® 450
Fournisseur: GERHARDT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12-0550 12-0540 12-0545
705-0860EA 13256 EUR
705-0860 705-0859 705-0861
Kjeldahl distillation systems, VAPODEST® 400 and VAPODEST® 450
Distillation Systems Kjeldahl Distillation Systems
The VAPODEST® 400 and 450 are designed with extended funtionality to allow easy, comfortable operation of Kjeldahl analysis.
  • 7" TFT colour display and easy menu navigation
  • Programmable, automatic addition of NaOH, H₂O and H₃BO₃
  • Programmable distillation and reaction times
  • Programmable steam power, 10 to 100% with steam soft start
  • Programmable suction of sample residue
  • Back lit glass components and easy clean surfaces
VAPODEST® 450 with inline connection for a optional titrator features: Automatic endpoint titration, display of consumed titration solution, display of pH value and data export.
Certifications: VAPODEST® 400 and 450 complie with the data management requirements of accredited laboratories (ISO 17025, GLP).